Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GC COLUMN HI-WAX20M 0.53MM 1.00UM 30M 1 * 1 items
Catalog # HIGCHI12-53-100-30
Supplier:
Avantor
Expand image
GC COLUMN HI-WAX20M 0.53MM 1.00UM 30M 1 * 1 items
Catalog # HIGCHI12-53-100-30
Supplier:
Avantor