Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
PH-PISTON 220µL 1 * 1 items
PH-PISTON 220µL 1 * 1 items
Print
PH-PISTON 220µL 1 * 1 items
Catalog # GILSF1077614
Supplier:
GILSON
PH-PISTON 220µL 1 * 1 items
Catalog # GILSF1077614
Supplier:
GILSON
Expand image
Return to Top
PH-PISTON 220µL 1 * 1 items
Add to cart
Price
11,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Return Policy
Product Details & Documents