0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Storage box for AFM discs 1 * 1 ST
PLAN16206
:
PLANO W. PLANNET
Storage box for AFM discs 1 * 1 ST
PLAN16206
:
PLANO W. PLANNET