ContactUsLinkComponent
Ammonium Standard Solution with 2,5 mg/L NH?-N for quality control checks with LCK505 Ammonium cuvette test. LCA505 is also suitable for quality checks with the Amtax sc Ammonium analyser. 1 * 1 items
Catalog # HACHLCA505
Supplier:  Hach
Ammonium Standard Solution with 2,5 mg/L NH?-N for quality control checks with LCK505 Ammonium cuvette test. LCA505 is also suitable for quality checks with the Amtax sc Ammonium analyser. 1 * 1 items
Catalog # HACHLCA505
Supplier:  Hach