Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GC COLUMN HI-17 0.53MM 2.00UM 10M 1 * 1 items
Catalog # HIGCHI07-53-200-10
Supplier:
Avantor
Expand image
GC COLUMN HI-17 0.53MM 2.00UM 10M 1 * 1 items
Catalog # HIGCHI07-53-200-10
Supplier:
Avantor