Order Entry
Export
Orders LinkContactUsLinkComponent
Inserts for U-D master manual samplers

Inserts for U-D master manual samplers

U-D Master insert split, Stainless steel 316L, 1 ml
Catalog # SAMP1230T-10S
Supplier:   Sampling Systems
Inserts for U-D master manual samplers
Catalog # SAMP1230T-10S
Supplier:  Sampling Systems
Inserts for U-D master manual samplers
Inserts for U-D master manual samplers

U-D Master insert split, Stainless steel 316L, 1 ml

Price
616,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Stainless steel inserts for U-D Master manual samplers.

  • Made from 316 stainless steel

These inserts are ideal for helping to prevent sampling error. By simply changing the sample inserts, different volumes can be accurately sampled.

Specifications
  • Volume:
    1 ml
  • Description:
    U-D Master insert split
  • Material:
    Stainless steel 316L
  • Seq:
    03