Lanosterol 50%
Glass bottle, 5 g
Catalog # ACRO466720050
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (3S,5R,10S,13R,14R,17R)-4,4,10,13,14-Pentamethyl-17-((R)-6-methylhept-5-en-2-yl)-2,3,4,5,6,7,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol, Cryptosterol
- CAS Number:79-63-0
- Formula:C₃₀H₅₀O
- MDL Number:MFCD00021108
- Molecular Weight:426.73 g/mol
- Melting Pt:137°C
- Storage Temperature:Ambient
- EINECS:201-214-9
Specifications
Specification Test Results
- Appearance:White to light yellow to green powder or crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥ 50.0%
- Specific optical rotation:+60.0° to +72.0° (20°C, 589nm) (c=1, Toluene)
Specifications
- Pack type:Glass bottle
- Pk:5 g



