Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]COLUMN AH 100 Å 3 µM 4.6 X 150 MM 1 * 1 ST
Catalog # SIELAH-46.150.0310
Supplier:
SIELC Technologies
Expand image
[EN]COLUMN AH 100 Å 3 µM 4.6 X 150 MM 1 * 1 ST
Catalog # SIELAH-46.150.0310
Supplier:
SIELC Technologies