Creatinine NF
Poly bottle, 25 g
Catalog # SPCMC1403-25GM
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
- CAS Number:60-27-5
- Formula:C₄H₇N₃O
- MDL Number:MFCD00059730
- Molecular Weight:113.12 g/mol
- Storage Temperature:Ambient
- EINECS:200-466-7
Specifications
Specification Test Results
- Assay:98.5% Min.
Specifications
- Pack type:Poly bottle
- Pk:25 g



