Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Tension infiltrometer set Tension infiltrometer for measuring hydraulic properties of unsaturated soil, complete standard set. 1 * 1 SET
Catalog # EIJK0909
Supplier:
EIJKELKAMP AGRISECTOR
Expand image
Tension infiltrometer set Tension infiltrometer for measuring hydraulic properties of unsaturated soil, complete standard set. 1 * 1 SET
Catalog # EIJK0909
Supplier:
EIJKELKAMP AGRISECTOR