Screw caps for SuperClear® microtubes
Blue
Catalog # 3600-877-000-9
Supplier: Labcon
Jump to:
Product Details & Documents
Caps only.
- Endotoxin Free (non-pyrogenic)
- Polyethylene USP Class VI resin
- Polypropylene
- Autoclavable
Specifications
- Colour:Blue
- Seq:02



