Order Entry
Export
Orders LinkContactUsLinkComponent
 

Screw caps for SuperClear® microtubes

Yellow
Catalog # 3600-872-000-9
Supplier:   Labcon
Screw caps for SuperClear® microtubes
Catalog # 3600-872-000-9
Supplier:  Labcon
Screw caps for SuperClear® microtubes
Screw caps for SuperClear® microtubes

Yellow

Price
156,00 €
Log in for availability
1 * 500 items
Product Details & Documents

Caps only.

  • Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Polypropylene
  • Autoclavable
Specifications
  • Colour:
    Yellow
  • Seq:
    09