Order Entry
Export
Orders LinkContactUsLinkComponent
Human Elastin ELISA Kit

Human Elastin ELISA Kit

Human Elastin ELISA kit, 96 tests
Catalog # ABCAAB239433-96
Supplier:   Abcam
Human Elastin ELISA Kit
Catalog # ABCAAB239433-96
Supplier:  Abcam
CAS Number:  
Human Elastin ELISA Kit
Human Elastin ELISA Kit

Human Elastin ELISA kit, 96 tests

CAS Number:  
Price
790,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human Elastin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Elastin with a sensitivity of 44 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.

Specifications

Specifications

  • Description:
    Human Elastin ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Citrate plasma, Serum, EDTA Plasma
  • Detection range:
    0,19 - 12 ng/ml
  • Storage temperature:
    +4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB239433-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    44 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Elastin
  • Regulatory status:
    RUO

Specifications