Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
ACBI1 1 * 5 mg
ACBI1 1 * 5 mg
Print
ACBI1 1 * 5 mg
Catalog # MDEXHY-128359-5MG
Supplier:
MEDCHEMEXPRESS
ACBI1 1 * 5 mg
Catalog # MDEXHY-128359-5MG
Supplier:
MEDCHEMEXPRESS
CAS Number:
Expand image
Return to Top
ACBI1 1 * 5 mg
Add to cart
CAS Number:
Price
283,00 €
Log in
for availability
Unit of Measure
1 * 5 mg
-
+
Add to cart
Return Policy
Product Details & Documents