Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
PH STRIPS PH 0 - 14 PLASTIC 1 * 100 items
PH STRIPS PH 0 - 14 PLASTIC 1 * 100 items
Print
PH STRIPS PH 0 - 14 PLASTIC 1 * 100 items
Catalog # LOVI56S001290
Supplier:
Lovibond
PH STRIPS PH 0 - 14 PLASTIC 1 * 100 items
Catalog # LOVI56S001290
Supplier:
Lovibond
Expand image
Return to Top
PH STRIPS PH 0 - 14 PLASTIC 1 * 100 items
Add to cart
Price
40,90 €
Log in
for availability
Unit of Measure
1 * 100 items
-
+
Add to cart
Return Policy
Product Details & Documents