D-(+)-Maltose monohydrate ≥92.0% (by anhydrous basis) NF
12 kg
Catalog # SPCMM1079-12KG
Supplier: Spectrum Chemical
D-(+)-Maltose monohydrate ≥92.0% (by anhydrous basis) NF
Catalog # SPCMM1079-12KG
Supplier: Spectrum Chemical
CAS Number: 6363-53-7
Jump to:
Product Details & Documents
Maltose, Monohydrate, NF is used in pharmaceutical formulations and as a parenteral supplement of sugar for diabetics. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
- CAS Number:6363-53-7
- Formula:C₁₂H₂₂O₁₁·1H₂O
- MDL Number:MFCD00149343
- Molecular Weight:360.32 g/mol
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay (Anhydrous Basis):92.0%
- Water Determination:4.5 - 6.5%
- pH:4.0 - 5.5
- Residue on Ignition:Max. 0.05%
- Dextrin, Starch and Sulfite:To Pass Test
- Identification:To Pass Test
- Residual Solvents:To Pass Test
- Elemental Impurities:As Reported
Specifications
- Pk:12 kg



