Order Entry
Export
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M526T

Biopsy processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M526T

Blue
Catalog # SIMPM526-6T
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M526T
Catalog # SIMPM526-6T
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M526T

Blue

Price
272,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents

These cassettes are made of acetal and are specially designed to hold biopsy specimens.

  • 1 mm square openings to maximise fluid exchange
  • Anterior writing area at a 45° angle
  • Taped cassettes to be used with Leica and Sakura ink jet printers
  • Easy to remove disposable cover

Unisette™ histology cassettes come with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.

1 mm opening maximises fluid exchange and ensures proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design of the UNISETTE™ prevents early separation of base and lid and allows for one-handed operation.

Packaging: Cassettes are packed in stacks of 40. Each case contains 25 stacks for a total of 1000 cassettes.

Specifications
  • Colour:
    Blue
  • Seq:
    05