Order Entry
Export
Orders LinkContactUsLinkComponent
 

Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T

Yellow
  SIMPM525-5T
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
  SIMPM525-5T
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T

Yellow

 
241,00 €
   
1 * 1.000 items
 

These cassettes are moulded from acetal and keep specimens safely submerged in liquid, and are resistant to the chemical action of most histological solvents.

  • Anterior writing area at a 45° angle
  • Taped cassettes to be used with Leica ink jet printers
  • Easy to remove disposable cover

Unisette™ histology cassette comes with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.

The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design of the UNISETTE™ prevents early separation of base and lid and allows for one-handed operation.

 : Cassettes are packed in stacks of 40. Each case contains 25 stacks for a total of 1000 cassettes.

 
  • Colour:
    Yellow
  • Seq:
    04