Orders LinkContactUsLinkComponent
Avantor® Hichrom HI-5 Basic, GC Columns
Avantor® Hichrom HI-5 Basic, GC Columns
GC column, Capillary, 0,50 µm, 0,25 mm, 30 m
Catalog # HI03-25-050-30
Supplier:   Avantor
Avantor® Hichrom HI-5 Basic, GC Columns
Catalog # HI03-25-050-30
Supplier:  Avantor
Avantor® Hichrom HI-5 Basic, GC Columns
Avantor® Hichrom HI-5 Basic, GC Columns
GC column, Capillary, 0,50 µm, 0,25 mm, 30 m
Price
723,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Max temperature limits may change depending on the film thickness.
Specifications
  • Column Format:
    Capillary
  • Film thickness:
    0,50 µm
  • Ø int.:
    0,25 mm
  • Description:
    GC column
  • Length:
    30 m
  • Cat. No.:
    HI03-25-050-30
  • Crossbond:
    Yes
  • Phase composition:
    5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • Polarity:
    Low-polarity stationary phase
  • Phase name:
    HI-5 Basic
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-100-PF-C
    Log in for availability
    1 * 100 items - 849,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-10-PF-C
    Log in for availability
    1 * 10 items - 117,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-10-PFY-C
    Log in for availability
    1 * 10 items - 1 020,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-1-PFY-C
    Log in for availability
    1 * 1 items - 105,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-25-PF-C
    Log in for availability
    1 * 25 items - 226,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-2-PFY-C
    Log in for availability
    1 * 2 items - 210,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-50-PF-C
    Log in for availability
    1 * 50 items - 435,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-5-PF-C
    Log in for availability
    1 * 5 items - 78,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-5-PFY-C
    Log in for availability
    1 * 5 items - 517,00 €