Orders LinkContactUsLinkComponent
Screw Caps for Bottles
Screw Caps for Bottles
Screw cap, tamper evident, with PTFE seal, 28 mm, White, HDPE
Catalog # RIXI8-1316-28-WS
Screw Caps for Bottles
Catalog # RIXI8-1316-28-WS
Screw Caps for Bottles
Screw Caps for Bottles
Screw cap, tamper evident, with PTFE seal, 28 mm, White, HDPE
Price
23,60 €
Log in for availability
1 * 25 items
Product Details & Documents
Specifications
  • Thread:
    28 mm
  • Colour:
    White
  • Description:
    Screw cap, tamper evident, with PTFE seal
  • Material:
    HDPE
  • Seq:
    29