Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CARTRIDGE T3 VANGUARD 120Å 2.7µM 2.1X5M 1 * 10 cm
Catalog # WATE186008506
Supplier:
WATERS
Expand image
CARTRIDGE T3 VANGUARD 120Å 2.7µM 2.1X5M 1 * 10 cm
Catalog # WATE186008506
Supplier:
WATERS