ContactUsLinkComponent
This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037) 1 * 25 µG
Catalog # NOVUNBP2-24729SS
Supplier:  Novus Biologicals
This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037) 1 * 25 µG
Catalog # NOVUNBP2-24729SS
Supplier:  Novus Biologicals