Hand held tool and covers
Hand held tool, 152×102 mm, PP
MICRHHT-06PP
The hand held tool conforms to the hand and works with a choice of M-Zone hand held mop covers to ergonomically clean and scrub surfaces for a complete sanitization of small equipment and production surfaces.
- Ergonomic cleaning for small equipment and surfaces
- Lightweight, forms to hand
- Available in polypropylene, recyclable plastic, or autoclavable polyketone adapters
- Irradiated NovaPoly™ Cover is 100% smooth knit polyester, matches traditional cleanroom wipe fabrics, and provides excellent disinfectant dispersal
- PolyMesh™ Cover is 100% polyester mesh, with foam interior, for scrubbing
- Covers compatible with alcohol and strong disinfectants and are launderable for reuse
- Autoclavable
The tool adapter is available in polypropylene, recyclable plastic, or an autoclavable polyketone. The slip-on covers are available in smooth polyester or polyester mesh fabric, to meet a variety of cleaning needs.
- Description:Hand held tool
- L×W:152×102 mm
- Material:PP
- Seq:01



