Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
N/400 PVSK SOLUTION
N/400 PVSK SOLUTION
Print
N/400 PVSK SOLUTION
Catalog # WAKO167-28105
Certificates
Supplier:
Wako
N/400 PVSK SOLUTION
Catalog # WAKO167-28105
Supplier:
Wako
CAS Number:
Certificates
Expand image
Return to Top
N/400 PVSK SOLUTION
Add to cart
CAS Number:
Price
56,70 €
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Add to cart
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number