Orders LinkContactUsLinkComponent
Cleaning cloths, Microfibre Light
White, 340×400 mm, 40 sheets per pack
  115-2497
Cleaning cloths, Microfibre Light
  115-2497
Cleaning cloths, Microfibre Light
Cleaning cloths, Microfibre Light
White, 340×400 mm, 40 sheets per pack
23,60 €
1 * 40 items
Microfibre Light cloths provide the same superior cleaning performance as traditional woven microfibre cloths without the risk of cross contamination. Designed for short term use.
  • Removes 99% of bacteria from surfaces, improving hygiene
  • Can be disposed of after use, reducing time spent by cleaning staff on non value added tasks
  • Unique structure of the Microfibre Light cloth ensures that bacteria are trapped in the cloth and not released onto the next surface
  • Can be used in food applications (FCC certificate)
Ideal for healthcare applications, food processing industries, building care and janitorial tasks.
  • Packed:
    40 sheets per pack
  • Colour:
    White
  • L×W:
    340×400 mm