Order Entry
Export
Orders LinkContactUsLinkComponent
Argatroban, Reference Standards

Argatroban, Reference Standards

Argatroban Related Compound B, USP reference standard, Vial, 20 mg
Catalog # 1042420.
Supplier:   USP
Argatroban, Reference Standards
Catalog # 1042420.
Supplier:  USP
CAS Number:  
Argatroban, Reference Standards

Argatroban Related Compound B, USP reference standard, Vial, 20 mg

CAS Number:  
Price
1 330,00 €
Log in for availability
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    20 mg
  • Standards:
    USP reference standard
  • Description:
    Argatroban Related Compound B
  • Cat. No.:
    1042420.
  • Description:
    Argatroban Related Compound B
  • Synonym(s):
    Ethyl (4R)-1-[N^8-nitro-L-arginyl]-4-methylpiperidine-2-carboxylate dihydrochloride

Specifications