Orders LinkContactUsLinkComponent
VWR® Maximum, Cut Resistant Gloves
11, Grey
Catalog # 112-5696
Supplier:   VWR Collection
VWR® Maximum, Cut Resistant Gloves
Catalog # 112-5696
Supplier:  VWR Collection
VWR® Maximum, Cut Resistant Gloves
VWR® Maximum, Cut Resistant Gloves
11, Grey
Price
75,60 €
Log in for availability
1 * 12 PAIR
Product Details & Documents
This VWR® glove with polyurethane coating provides superior cut protection for applications that include a high risk of cuts.
  • Nylon/spandex/polyester blend composition
  • Polyurethane (PU) coating on palm and fingertips
  • Grey knitted glove with grey coating
Ideal for handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance, and janitorial applications.
 : Meets requirements for EN 388:2016 (4X42E). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications
  • Glove size:
    11
  • Colour:
    Grey
  • Seq:
    06
  • Cat. No.:
    112-5696
  • Latex / latex-free:
    Latex-free
  • Disposable / reusable:
    Reusable
  • Cuff style:
    Knit wrist
  • EN norm:
    EN 388:2016 (4X42E)
  • Finish:
    PU palm coating
  • Ambidextrous / hand-specific:
    Hand-specific
  • Gauge:
    13