Nailsaver scouring sponges
Red/white, 133×70 mm
Catalog # MMMASCH106
Supplier: 3M
Jump to:
Product Details & Documents
3M™ Nailsaver scouring sponges are cleaning sponges made of a textured synthetic fibre white web containing very soft abrasive particles, laminated with an adhesive onto a grooved synthetic colour-coded foam holder.
- Easy positioning four colours of foams) to comply with HACCP requirements
- Good cleaning performance for light-duty cleaning tasks
- Soft and low scratching power on most surfaces
- Easy to handle and finger protection thanks to the shape of the foam
- Very long lasting
For easy cleaning with low scratching power on most surfaces, use 3M™ Nailsaver scouring sponges.
These scouring sponges offer good cleaning performance for light-duty cleaning tasks with soft and low scratching power on most surfaces. They’re very long lasting and easy to handle thanks to the shape of the foam.
Specifications
- Colour:Red/white
- L×W:133×70 mm
- Seq:03