L(+)-Valine USP
20 kg
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: H-Val-OH, Valine, (S)-(+)-Valine
- CAS Number:72-18-4
- Formula:C₅H₁₁NO₂
- MDL Number:MFCD00064220
- Molecular Weight:117.15 g/mol
- Merck Index:13,09458
- Boiling Pt:217 °C (1013 hPa)
- Melting Pt:86 °C
- Density:1.284 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-773-6
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay:98.5% Min.
Specifications
- Pk:20 kg



