ContactUsLinkComponent
Recombinat Protein,Human CD300c LMIR2 Protein, His Tag,Human CD300c, His Tag (CDC-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Arg 183 (Accession AAH22279).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBCDC-H5224-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human CD300c LMIR2 Protein, His Tag,Human CD300c, His Tag (CDC-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Arg 183 (Accession AAH22279).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBCDC-H5224-1MG
Supplier:  ACROBIOSYSTEMS