ContactUsLinkComponent
Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630  (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The protein 1 * 100 µG
Catalog # ACRBCD8-H5247100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630  (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The protein 1 * 100 µG
Catalog # ACRBCD8-H5247100UG
Supplier:  ACROBIOSYSTEMS