ContactUsLinkComponent
Recombinat Protein,Human C-Reactive Protein (CRP) Protein, His Tag,Human C-Reactive Protein, His Tag (CRP-H5226) is expressed from h uman 293 cells (HEK293). It contains AA Gln 19 - Pro 224 (Accession NP000558.2).This protein carries a polyhistidine 1 * 1 mg
Catalog # ACRBCRP-H5226-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human C-Reactive Protein (CRP) Protein, His Tag,Human C-Reactive Protein, His Tag (CRP-H5226) is expressed from h uman 293 cells (HEK293). It contains AA Gln 19 - Pro 224 (Accession NP000558.2).This protein carries a polyhistidine 1 * 1 mg
Catalog # ACRBCRP-H5226-1MG
Supplier:  ACRO Biosystems