ContactUsLinkComponent
Recombinat Protein,Human Cystatin SA CST2 Protein, His Tag,Human Cystatin SA, His Tag (CS2-H5224) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Ala 141 (Accession AAH62679).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
Catalog # ACRBCS2-H5224-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Cystatin SA CST2 Protein, His Tag,Human Cystatin SA, His Tag (CS2-H5224) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Ala 141 (Accession AAH62679).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
Catalog # ACRBCS2-H5224-1MG
Supplier:  ACROBIOSYSTEMS