ContactUsLinkComponent
Recombinat Protein,Human DLL4 Protein, His Tag,Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contai ns AA Ser 27 - Pro 524 (Accession AAI06951).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 1 mg
Catalog # ACRBDL4-H5227-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human DLL4 Protein, His Tag,Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contai ns AA Ser 27 - Pro 524 (Accession AAI06951).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 1 mg
Catalog # ACRBDL4-H5227-1MG
Supplier:  ACROBIOSYSTEMS