ContactUsLinkComponent
Recombinat Protein,Human S100P S100E Protein, Tag Free,Human S100P, Tag Free (S1P-H5110) is expressed from E.coli cells. It contains AA Met 1 - Lys 95 (Accession AAH06819).This protein carries no tag. The protein has a calculated MW of 10.4 kDa. The 1 * 1 mg
Catalog # ACRBS1P-H5110-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human S100P S100E Protein, Tag Free,Human S100P, Tag Free (S1P-H5110) is expressed from E.coli cells. It contains AA Met 1 - Lys 95 (Accession AAH06819).This protein carries no tag. The protein has a calculated MW of 10.4 kDa. The 1 * 1 mg
Catalog # ACRBS1P-H5110-1MG
Supplier:  ACROBIOSYSTEMS