ContactUsLinkComponent
Recombinat Protein,Human uPAR PLAUR Protein, His Tag,Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession AAH02788).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBUPR-H5226-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human uPAR PLAUR Protein, His Tag,Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession AAH02788).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBUPR-H5226-1MG
Supplier:  ACROBIOSYSTEMS