ContactUsLinkComponent
Recombinat Protein,Human Serpin A3 Protein, His Tag,Human Serpin A3, His Tag (SE3-H5221) is expressed from human 293 cells (HEK293). It contains AA His 24 - Ala 423 (Accession AAH03559).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBSE3-H5221-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Serpin A3 Protein, His Tag,Human Serpin A3, His Tag (SE3-H5221) is expressed from human 293 cells (HEK293). It contains AA His 24 - Ala 423 (Accession AAH03559).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBSE3-H5221-1MG
Supplier:  ACRO Biosystems