Human recombinant CD229 (from HEK293)
100 µG
ACRBCD9-H52H6100UG
- Pk:100 µG
- Protease-free:Y
- Protein/peptide name:CD229
- Protein synonyms:LY9|hly9|Ly-9|SLAMF3|Ly9
- Purity:>90%
- Endotoxin level:Low
- Source:HEK293 cells
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Molecular weight:46,7 kDa
- Shipping temperature:Powder, RT
- Nuclease-free:Y
- Cat. No.:ACRBCD9-H52H6100UG
- Protein/peptide type:Recombinant
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Storage conditions:–20 °C
- Gene ID:Q9HBG7-1
- Carrier-free:Y
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Conjugation:Unconjugated
- Species:Human
- Animal-free:Y



