ContactUsLinkComponent
Recombinat Protein,Human NKG2D CD314 Protein, His Tag,Human NKG2D, His Tag (NKD-H5245) is expressed from human 293 cells (HEK293). I t contains AA Ile 73 - Val 216 (Accession P26718-1).This protein carries a polyhistidine tag at the N-terminus. The p 1 * 100 µG
Catalog # ACRBNKD-H5245100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human NKG2D CD314 Protein, His Tag,Human NKG2D, His Tag (NKD-H5245) is expressed from human 293 cells (HEK293). I t contains AA Ile 73 - Val 216 (Accession P26718-1).This protein carries a polyhistidine tag at the N-terminus. The p 1 * 100 µG
Catalog # ACRBNKD-H5245100UG
Supplier:  ACROBIOSYSTEMS