Orders LinkContactUsLinkComponent
PC certified clean validation bottles, Nalgene®
PC certified clean validation bottles, Nalgene®
20 ml, PC certified clean validation bottles
Catalog # 216-2288
   Low Manufacturing Impact
PC certified clean validation bottles, Nalgene®
Catalog # 216-2288
   Low Manufacturing Impact
PC certified clean validation bottles, Nalgene®
PC certified clean validation bottles, Nalgene®
20 ml, PC certified clean validation bottles
Price
3 810,00 €
Log in for availability
1 * 504 items
Product Details & Documents
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Polycarbonate construction
  • Sterile
  • Clear finish
  • Certified clean
These bottles can reduce the risk of contamination.
Certifications: Validated to USP 788 and <0,5 EU/ml endotoxin.
Specifications
  • Capacity:
    20 ml
  • Description:
    PC certified clean validation bottles
  • Seq:
    02
  • Cat. No.:
    216-2288
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact