Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EPA Method 8260 VOC Mixture 3 2000 µg/mL in Methanol 1 * 1 KIT
Catalog # EHERYS09000010ME
Supplier:
EHRENSTORFER
Expand image
EPA Method 8260 VOC Mixture 3 2000 µg/mL in Methanol 1 * 1 KIT
Catalog # EHERYS09000010ME
Supplier:
EHRENSTORFER