Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMN NUCLEOSIL 100 5UM SA 50X4.6MM EC 1 * 1 items
Catalog # MANA720793.46
Supplier:
MACHEREY-NAGEL
Expand image
COLUMN NUCLEOSIL 100 5UM SA 50X4.6MM EC 1 * 1 items
Catalog # MANA720793.46
Supplier:
MACHEREY-NAGEL