ContactUsLinkComponent
CAROUSEL HOLDER 5 POSITION 1'' DR6000 Cell holder with 5 positions to measure mini series or enzymatic methods. 1 * 1 items
Catalog # HACHLZV9029900011
Supplier:  Hach
CAROUSEL HOLDER 5 POSITION 1'' DR6000 Cell holder with 5 positions to measure mini series or enzymatic methods. 1 * 1 items
Catalog # HACHLZV9029900011
Supplier:  Hach