Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Troemner OIML Precision Weights Weight, 20 kg, OIML E2, NVLAP Cert 1 * 1 items
Catalog # OHAU80780306
Supplier:
OHAUS
Expand image
Troemner OIML Precision Weights Weight, 20 kg, OIML E2, NVLAP Cert 1 * 1 items
Catalog # OHAU80780306
Supplier:
OHAUS