Sheets for ÄKTA Systems, Benchkote®, Whatman™
Benchkote® sheet for ÄKTA go, White
Catalog # WHAT2300-10092
Supplier: Whatman products (Cytiva)
Sheets for ÄKTA Systems, Benchkote®, Whatman™
Catalog # WHAT2300-10092
Supplier: Whatman products (Cytiva)
Jump to:
Product Details & Documents
These Benchkote® sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
- Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems
Benchkote® material has a high quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
Specifications
- Colour:White
- Description:Benchkote® sheet for ÄKTA go