Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT CANINE IL-31 PROTEIN 1 * 50 µG
Catalog # NOVUNBP2-59591
Supplier:
Novus Biologicals
Expand image
RECOMBINANT CANINE IL-31 PROTEIN 1 * 50 µG
Catalog # NOVUNBP2-59591
Supplier:
Novus Biologicals