Single-channel pipettes, mechanical, fixed / variable volume, Transferpette® S
Micro starter kit incl. 3× pipettes (0,1-2,5 µl, 0,5-10 µl, 10-100 µl), 3× shelf mounts
613-6734
: Brand
Single-channel pipettes, mechanical, fixed / variable volume, Transferpette® S
613-6734
: Brand
The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Due to its perfect ergonomics, the Transferpette® S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands.
- True one-hand operation for right- and left-handed users: Easily set the volume, pipette and eject the tip
- Colour code on volume adjustment wheel, shaft and volume-change protection
- Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
- Slim shaft for sampling from narrow vessels (centrifuge tubes)
- Universal tip cone
- Fully autoclavable at 121 °C
The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette® S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
- Description:Micro starter kit incl. 3× pipettes (0,1-2,5 µl, 0,5-10 µl, 10-100 µl), 3× shelf mounts
- Seq:29
- Cat. No.:613-6734
- Environmentally Preferable:Low Manufacturing Impact|Sustainable Packaging
Accessories
- DescriptionFor
- BRND704807Transferpette® S Carousel Benchtop Stand2020 and later Transferpette® S pipettes1 * 1 items - 65,10 €
- DescriptionFor
- BRND704811Transferpette® S Shelf Mount2020 and later Transferpette® S pipettes1 * 1 items - 11,80 €
- DescriptionFor
- BRND704812Transferpette® S Wall Mount2020 and later Transferpette® S pipettes1 * 1 items - 18,80 €