Orders LinkContactUsLinkComponent
Axygen® multirack maximum Recovery® tips
Standard Length, +
  AXYGMR-300-L-R-S
 :   Corning
Axygen® multirack maximum Recovery® tips
  AXYGMR-300-L-R-S
 :  Corning
Axygen® multirack maximum Recovery® tips
Axygen® multirack maximum Recovery® tips
Standard Length, +
469,00 €
1 * 4.800 items
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
Axygen® MultiRack Maximum Recovery® tips are available with or without aerosol filters. Axygen® Maximum Recovery® technology creates a hydrophobic surface to improve accuracy and precision while reducing surface tension and sample wastage. They are specially designed for lab applications that demand complete release of a liquid solution, such as low concentration DNA, selected protein, and viscous liquids (glycerin, Triton-X and more). Axygen® MultiRack Maximum Recovery® tips are available in a wide range of tip sizes with filters and sterilization.
 : Axygen® MultiRack pipette tips are also available in bulk, hinged rack, filter tips, reload, and tipstation configurations.
  • Sterile:
    +
  • Seq:
    02
  • Tip style:
    Standard Length