Orders LinkContactUsLinkComponent
MultiRack reloads, Axygen®
Extended length, 0,1 - 10 µl, Reload: 960 Tips/Pack, 4 Packs/Case
Catalog # AXYGMR-10XT-RL
Supplier:   Corning
MultiRack reloads, Axygen®
Catalog # AXYGMR-10XT-RL
Supplier:  Corning
MultiRack reloads, Axygen®
MultiRack reloads, Axygen®
Extended length, 0,1 - 10 µl, Reload: 960 Tips/Pack, 4 Packs/Case
Price
185,00 €
Log in for availability
1 * 3.840 items
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack reload system is designed to fit Axygen® MultiRack as well as racks from other suppliers without tedious realignment. This design not only saves lab space but also uses less plastic to reduce environmental impact. Axygen® MultiRack reload tips are available in a wide range of sizes (10 µl extended, 200, 300 and 1000 µl extended).
Specifications
  • Packed:
    Reload: 960 Tips/Pack, 4 Packs/Case
  • Volume:
    0,1 - 10 µl
  • Seq:
    01
  • Tip style:
    Extended length