Glutaraldehyde 25% in aqueous solution for electron microscopy
2,5 L
: Glutaral, Glutardialdehyde, 1,5-Pentanedial
- :111-30-8
- :C₅H₈O₂
- :MFCD00007025
- :100.12 g/mol
- :–6 °C
- :Ambient
- :2922
- :8 (6.1),II
- :Density: 1.014 g/cm³ (20 °C)
- Appearance:clear colorless or yellow liquid
- Assay :23,5 – 26,5%
- Color scale:max. 50 APHA
- pH:min. 3,0
- Density (d20°C/4°C):1,060 - 1,065
- Absorbance A (235 nm; 1:200 diluted with water; 1 cm):max. 0,6
- Pk:2,5 L



